Learn More
Pritelivir, MedChemExpress
Product Code. 16402547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16402547
Brand: Medchem Express HY15303
MedChemExpress Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.
Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.
Specifications
Chemical Name or Material Pritelivir
Physical Form Powder
CAS 348086-71-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H18N4O3S2
For Use With (Application) COVID-19-anti-virus
Synonym AIC316 BAY 57-1293
Solubility Information DMSO : ≥ 33 mg/mL (81.99 mM)
SMILES O=C(N(C1=NC(C)=C(S(=O)(N)=O)S1)C)CC2=CC=C(C3=NC=CC=C3)C=C2
Molecular Weight (g/mol) 402.49
Formula Weight 402.49
Percent Purity 98.02%
Grade Research