Learn More
Invitrogen™ MOZ Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571427
This item is not returnable.
View return policy
Description
MOZ Polyclonal Antibody for Western Blot
Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.
Specifications
| MOZ | |
| Polyclonal | |
| Unconjugated | |
| Kat6a | |
| 1500036M03; 9930021N24Rik; histone acetyltransferase KAT6A; histone acetyltransferase MYST3; K(lysine) acetyltransferase 6A; KAT6A; lysine acetyltransferase 6A; monocytic leukemia zinc finger homolog; Monocytic leukemia zinc finger protein; Moz; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3; MRD32; MYST histone acetyltransferase (monocytic leukemia) 3; Myst3; MYST-3; runt-related transcription factor binding protein 2; Runt-related transcription factor-binding protein 2; RUNXBP2; ZC2HC6A; Zfp220; zinc finger protein 220; ZNF220 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7994 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92794 | |
| Kat6a | |
| synthetic peptide directed towards the N terminal region of human MOZ. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |