Learn More
Logistik™ Howie-Style Unisex Bottle Green Laboratory Coats with Knitted Cuffs
Product Code. 12328647
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Size (Metric) Chest:
101.6 to 104.14 cm
106.68 to 111.76 cm
116.84 to 121.92 cm
127 to 132.08 cm
91.44 to 96.52 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Size (Metric) Chest unitSize
12328647 Medium 101.6 to 104.14 cm Each
12851348 Small 91.44 to 96.52 cm Each
12831348 Large 106.68 to 111.76 cm Each
12861348 X-Large 116.84 to 121.92 cm Each
12811348 2X-Large 127 to 132.08 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12328647 Supplier Logistik™ Supplier No. 441YHV6BG100C
Only null left
Add to basket
Add to basket
Designed to conform to Howie report guidelines, side fastening with four layers of fabric at the front for extra protection against minor chemical splashes. Logistik™ Howie-Style Unisex Bottle Green Laboratory Coats with Knitted Cuffs are a high quality option for protection in the lab.
  • High-quality Howie style laboratory coat with knitted cuffs
  • With two lower pockets and wrapover style
  • Stud fastened front
  • Polyester/cotton
  • Not CE marked
Specifications
Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Cotton, Polyester
Color Bottle Green
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Style Howie
No. of Pockets 2
Collar Type Round
Size (Metric) Chest 101.6 to 104.14 cm
Includes With two lower pockets and wrap over style stud fastened front
Length (Metric) 100 cm
Construction Polyester, Cotton