Learn More
Masterflex™ L/S™ Standard Pump Head for Precision Tubing L/S™ PPS Housing, SS Rotor
Click to view available options
Tubing Size:
14
15
16
17
24
Unit Size:
Each
Description
Ideal for fluid transfer applications requiring high accuracy and repeatability
- Standard heads are precision molded to accommodate a single size of tubing
- Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
- Standard heads are compact and easy to integrate into your OEM applications
- Stack up to four heads depending on drive recommendations
Specifications
Specifications
| Dimensions (L x W x H) | 7.6 H x 7 W x 6.4 cm D |
| Flow Rate | 2.8 to 1700 mL/min. |
| For Use With (Application) | For fluid transfer applications requiring high accuracy and repeatability |
| Includes | Tubing loading key, single-channel mounting hardware, and a 15 in. (38cm) length of Silicone pump tubing |
| No. of Channels | 1 |
| No. of Rollers | 3 |
| Occlusion Type | Fixed |
| Overall Depth (Metric) | 6.4 cm |
| Overall Height (Metric) | 7.6 cm |
| Overall Width (Metric) | 7 cm |
| Show More |