Learn More
Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Product Code. 11363548
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11363548 25 g 25g
11373548 100 g 100g
2 options
Product Code. 11363548
Brand: Thermo Scientific Alfa Aesar 039174.14
Request Bulk or Custom Format
Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2233-42-3
Molecular Formula C32H60O8Zr
Molecular Weight (g/mol) 664.048
MDL Number MFCD00072684
InChI Key OFYFURKXMHQOGG-UHFFFAOYSA-J
Synonym zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate
PubChem CID 6451377
IUPAC Name 2-ethylhexanoate;zirconium(4+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4]
Specifications
Color Amber
Flash Point >93°C (199°F)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 664.04
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material Zirconium(IV) 2-ethylhexanoate
RUO – Research Use Only