Learn More
Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/B
Product Code. 11310574
Change view
Click to view available options
Diameter (Metric):
21 mm
24 mm
25 mm
37 mm
42.5 mm
47 mm
55 mm
70 mm
90 mm
110 mm
125 mm
150 mm
185 mm
Unit Size:
Pack of 100
Pack of 25
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11310574 37 mm Pack of 100
11390354 21 mm Pack of 100
11390564 24 mm Pack of 100
11300574 25 mm Pack of 100
11320574 42.5 mm Pack of 100
11330574 47 mm Pack of 100
10129710 55 mm Pack of 100
11350574 70 mm Pack of 100
11360574 90 mm Pack of 25
11370574 110 mm Pack of 25
11380574 125 mm Pack of 25
11390574 150 mm Pack of 25
11300584 185 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11310574 Supplier Cytiva Supplier No. 1821037
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B are particularly useful for liquid clarification and liquid scintillation counting.
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B Sheets are particularly useful for liquid clarification and liquid scintillation counting.
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • Fine particle filter with high wet-strength and high loading capacity
  • Useful for liquid clarification and liquid scintillation counting
  • Particularly useful when liquid clarification or solids quantification is required for heavily loaded fine particulate suspensions
  • Used in LSC techniques where high loading capacity is required
  • Can be used as a finely retentive membrane filter
TRUSTED_SUSTAINABILITY
Specifications
Basis Weight 143 g/m2
Description GE’s Whatman brand Grade GF/B is a thick filter with fine particle filter and good loading capacity. GF/B filters are particularly useful for liquid clarification and liquid scintillation counting.
Format Disc
Material Borosilicate Glass
Particle Retention 1.0 μm
Flow Rate 5.5 sec./100mL
For Use With (Application) Used in LSC techniques where high loading capacity is required.
Grade Grade GF/B
Quantity 100 Pack
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.02 in.
Thickness (Metric) 675 μm
Type Filter
Waterflow Rate 81 mL/min
Diameter (English) 1.4 in.
Diameter (Metric) 37 mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • FILTER GLASS CIRCLE
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.