Learn More
Merck Diffusion Chamber Rings
Product Code. 10629131 Shop All Merck Products
Change view
Click to view available options
Includes:
0.59 mm hole
No hole
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10629131 0.59 mm hole Pack of 50
10035291 No hole Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10629131 Supplier Merck Supplier No. PR0001401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Plexiglas rings with 14mm dia. for use with Diffusion chamber
  • Available with or without hole
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Diffusion Chamber Kit
Material Plexiglas™
Includes 0.59 mm hole
Type Plexiglas™ Rings
Diameter (English) 0.55 in.
Diameter (Metric) 14 mm