Learn More
VGF Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26781-1-AP-20UL
This item is not returnable.
View return policy
Description
The VGF antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human VGF. This antibody recognizes human, mouse antigen. The VGF antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| VGF | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 7425 | |
| IgG |
| Western Blot, Immunofluorescence | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |