Learn More
Vesicular Stomatitis Indiana Virus Glycoprotein Antibody [Janelia Fluor™ 635], R&D Systems™
Product Code. 30163585
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30163585 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30163585 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP326070JF635
 Restricted Item
Estimated Shipment: 11-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
Vesicular Stomatitis Indiana Virus Glycoprotein Polyclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen Vesicular Stomatitis Indiana Virus Glycoprotein
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein (Accession#: NP_041715.1; Uniprot#: P03522; Met1-Ser467).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus