Description
- VertiKlean MAX Hands-Free Mop Removal Tool.
- For Compact Bucket System.
Specifications
Specifications
| Product Type | Mop Head Removal Tool, Waste Bag Holder |
| Color Code | White |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | Compact Bucket System |
| Description | VertiKlean MAX Hands-Free Mop Removal Tool |
| Color-coded | No |
| Material | Stainless Steel |