Learn More
Saint Gobain Life Sciences Versilic™ Folding Skirt Stoppers
Product Code. 11744608
Change view
Click to view available options
Diameter (Metric) Inner:
12.1 to 15.6 mm
17.7 to 22.1 mm
20.9 to 26.4 mm
27.6 to 36.3 mm
9.8 to 13.2 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11744608 27.6 to 36.3 mm Pack of 5
11754598 9.8 to 13.2 mm Pack of 50
11764598 12.1 to 15.6 mm Pack of 50
11724608 17.7 to 22.1 mm Pack of 20
11734608 20.9 to 26.4 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11744608 Supplier Saint Gobain Life Sciences Supplier No. 46703005
  • ONLINE EXCLUSIVE PRICE
    421.74 NOK
    280.00 NOK / Pack of 5
    Save 141.74 NOK
    33% Off
Estimated Shipment: 15-12-2026
Silicone biologically neutral Versilic™ Folding Skirt Stoppers.
  • Provides complete tightness on glass, metal or plastic containers regardless of their rims.
  • The turnover is molded to grip the outside of the container neck, providing a double seal in conjunction with the annular serrations on the hollow plug.
  • Can be perforated.
  • Remains watertight after several perforations.
  • Recommended for biological applications.
  • Temperatures between -60°C and +220°C.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 27.6 to 36.3 mm
Quantity 5 Pk.
Temperature Range (Metric) -60°C to +200°C
Material Silicone
Product Type Silicone Stopper