Learn More
Ansell™ VersaTouch™ 92-200 Series Single-Use Blue Nitrile Gloves
Product Code. 12657051 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
Product Code. Size unitSize
12657051 10 Pack of 100
12627051 7 Pack of 100
12637051 8 Pack of 100
12647051 9 Pack of 100
4 options
Product Code. 12657051
Brand: Ansell™ 92200100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling.
  • Latex-free: no risk of Type 1 allergies.
  • Approved for food contact applications.
  • Ideal for handling all fatty food.
TRUSTED_SUSTAINABILITY
Specifications
Exterior Finish Fully Textured
Material Nitrile
Size 10
Thickness (English) 2.9 mil
Thickness (Metric) 0.07 mm
Model 9220010
Series 92-200