Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 450 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
Brand:  Cytiva (formerly Pall Lab) 66410
Additional Details : Weight : 0.31350kg
 View more versions of this product
Product Code. 17104571
  • 4180.00 NOK / Pack of 25
Estimated Shipment: 19-09-2024
Description
  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Versapor (Acrylic Copolymer on a Non-woven Support)
Non-sterile
142 mm
0.45 μm
94 μm