Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
Click to view available options
Product Type:
Hood with Faceseal
Hood with Inner Shroud
Hood with Neck Collar
Painter's Hood with Inner Shroud Assembly
Respirator Hood
Color:
Gray
White
Model:
S-433 L
S-433 L-5
S-433 S
S-605-10
S-607-10
S-655
S-657
S-707-10
S-757
S-805-5
S-855E
Unit Size:
Each
Pack of 10
Pack of 5
Description
- S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Provides head, face, neck and shoulder coverage from dust and paint spray.
- Provides a good field of view.
- Helps protect eyes and face from liquid splash and low energy flying particles and dust.
- Versatile accessories available that can help you adapt the hood to your particular environment.
Specifications
Specifications
| Assigned Protection Factor (APF) | 1000 |
| Certifications/Compliance | NIOSH |
| Description | Painter's Hood, S-757, with Inner Shroud, S-707 |
| Color | White |
| Material | Polypropylene |
| Product Type | Hood with Inner Shroud |
| For Use With (Equipment) | 3M™ premium head suspension of 3M™ powered air purifying and supplied air respirator system |
| Includes | Replacement Painter's Hood with Inner Shroud |
| Material (Lens) | PETG |
| Model | S-707-10 |
| Show More |