Learn More
Vericiguat, MedChemExpress
Product Code. 16487176
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487176 100 mg 100mg
16479717 5 mg 5mg
16477188 10 mg 10mg
16440286 2 mg 2mg
16403217 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16487176 Supplier Medchem Express Supplier No. HY16774
MedChemExpress Vericiguat (BAY1021189) is a potent, orally available and soluble guanylate cyclase stimulator.
Vericiguat (BAY1021189) is a potent, orally available and soluble guanylate cyclase stimulator.
Specifications
Chemical Name or Material Vericiguat
CAS 1350653-20-1
Color Light Brown
Purity Grade Notes Research
Molecular Formula C19H16F2N8O2
Quantity 100 mg
Synonym BAY1021189
Solubility Information DMSO : 60 mg/mL (140.72 mM; Need ultrasonic)
SMILES O=C(OC)NC1=C(N)N=C(C2=NN(CC3=CC=CC=C3F)C4=NC=C(F)C=C42)N=C1N
Molecular Weight (g/mol) 426.38
Formula Weight 426.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation