Learn More
Phenomenex™ Verex™ Screw Caps with Septa, 9 mm, Pre-Slit (PFAS-Free)
Product Code. 31745611
Change view
Click to view available options
Quantity:
100 Pk.
1000 Pk.
Unit Size:
Pack of 100
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31745611 100 Pk. Pack of 100
31748602 1000 Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31745611 Supplier Phenomenex™ Supplier No. AR089P712
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Accessories for use with chromatography vials and in various chromatography applications.
  • Analysis: Cleanliness and performance tested as standard
  • Certification: 100% defect-free
  • Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Diameter (Metric) Outer 9 mm
Material Polypropylene
Pre-slit No
Sterility Sterile
Septum Silicone/Polypropylene
Quantity 100 Pk.
Closure Size 9 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color Blue/Clear