Learn More
Phenomenex™ Verex™ Preassembled Screw Caps, 9 mm, with Septa, Pre-Slit
Product Code. 31744388
Change view
Click to view available options
Color:
Black
Blue
Septum:
PTFE/Rubber
PTFE/Silicone
Quantity:
100 Pk.
1000 Pk.
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Septum Quantity unitSize
31744388 Blue PTFE/Silicone 100 Pk. Pack of 100
31747078 Blue PTFE/Silicone 1000 Pk. Pack of 1000
31743300 Black PTFE/Silicone 100 Pk. Pack of 100
31748819 Black PTFE/Silicone 1000 Pk. Pack of 1000
31747080 Blue PTFE/Rubber 100 Pk. Pack of 100
31741749 Blue PTFE/Rubber 1000 Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31744388 Supplier Phenomenex™ Supplier No. AR0897212
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Accessories for use with chromatography vials and in various chromatography applications.
  • Analysis: Cleanliness and performance tested as standard
  • Certification: 100% defect-free
  • Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Diameter (Metric) Outer 9 mm
Pre-slit Yes
Sterility Sterile
Septum PTFE/Silicone
Quantity 100 Pk.
Closure Size 9 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap