Description
- Suitable for Buerkle™ storage bottles, with or without threaded connector
- For use with 5 L, 10 L, 25 L, and 60 L storage bottles
Specifications
Specifications
| Color | White, Red |
| Material | Polypropylene, Polyethylene |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| For Use With (Equipment) | 5 L, 10 L, 25 L, 60 L Storage Bottles |
| Closure Type | Vented |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Product Type | Ventilation Seal |