Learn More
Veliparib dihydrochloride, MedChemExpress
Product Code. p-200100092
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16484576
Brand: Medchem Express HY10130
MedChemExpress Veliparib (dihydrochloride) is a potent inhibitor of PARP1 and PARP2 with Kis of 5.2 nM and 2.9 nM in cell-free assays, respectively.
Veliparib (dihydrochloride) is a potent inhibitor of PARP1 and PARP2 with Kis of 5.2 nM and 2.9 nM in cell-free assays, respectively.
Specifications
Chemical Name or Material Veliparib dihydrochloride
Physical Form Solid
CAS 912445-05-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H18Cl2N4O
For Use With (Application) Cancer-programmed cell death
Synonym ABT-888 dihydrochloride
Solubility Information H2O : 250 mg/mL (788.12 mM; Need ultrasonic) ∣DMSO : ≥ 3.2 mg/mL (10.09 mM)
SMILES O=C(C1=C2NC([C@@]3(NCCC3)C)=NC2=CC=C1)N.Cl.Cl
Molecular Weight (g/mol) 317.21
Formula Weight 317.21
Percent Purity 99.27%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.