missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VEGFD Polyclonal Antibody
GREENER_CHOICE

Product Code. 13404107
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
13404107 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 13404107

Brand: Invitrogen™ PA529806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Predicted reactivity: Mouse (91%), Rat (87%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGFD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene VEGFD
Gene Accession No. O43915
Gene Alias AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand
Gene Symbols VEGFD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VEGFD. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.