Description
The VEGFA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human VEGFA. This antibody recognizes human antigen. The VEGFA antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | VEGFA |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Co-immunoprecipitation (Co-IP), ELISA |
| Classification | Polyclonal |
| Concentration | 233 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 7422 |
| Target Species | Human |
| Show More |