Learn More
VEGFA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26381-1-AP-20UL
This item is not returnable.
View return policy
Description
The VEGFA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human VEGFA. This antibody recognizes human antigen. The VEGFA antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
| VEGFA | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 7422 | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, Co-immunoprecipitation (Co-IP), ELISA | |
| 233 μg/mL | |
| Rabbit | |
| Primary | |
| Human |