missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
VEGF Monoclonal specifically detects VEGF in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
| Dilution | Immunohistochemistry-Paraffin 2 - 4 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml |
| Gene Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?