missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (123413) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9381JF669
5095.75 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal antibody specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| VE-Cadherin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin, Asp48-Gln593, Accession # P33151 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 123413 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction