Learn More
VD3-d6, MedChemExpress
Description
VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3
Specifications
Specifications
| Chemical Name or Material | VD3-d6 |
| CAS | 118584-54-6 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C27H38D6O |
| Color | Light Yellow |
| Synonym | Vitamin D3-26,26,26,27,27,27-d6 |
| Solubility Information | DMSO : 100 mg/mL (255.97 mM; Need ultrasonic) |
| SMILES | O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2 |
| Industry Type | Neuroscience-Neuromodulation |
| Show More |