Learn More
Sonics Materials VCX 750 Ultrasonic Microprocessor
Product Code. 11763619
Change view
Click to view available options
Wattage:
750 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage Unit Size
11763619 750 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11763619 Supplier Sonics Materials Supplier No. VCX750220
  • ONLINE EXCLUSIVE PRICE
    63135.00 NOK
    58786.15 NOK / Pack of 1
    Save 4348.85 NOK
    6% Off
 Restricted Item
Estimated Shipment: 25-11-2026
The 750 Watt Ultrasonic Processor is for small and medium volume applications and is ideal for safely processing a wide range of organic and inorganic materials from 250 microliters to 1 liter.
  • Ultrasonic processors for small and medium volume applications
  • 500 and 750 watt ultrasonic processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
  • Energy monitor, digital wattmeter, microprocessor-based and programmable
  • 10 hour process timer
Specifications
Wattage 750 W
For Use With (Application) Small and medium volume applications
Timer 10 hr. process timer