Learn More
Sonics Materials VCX 500 Ultrasonic Microprocessor
Product Code. 11753619
Change view
Click to view available options
Wattage:
500 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage Unit Size
11753619 500 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11753619 Supplier Sonics Materials Supplier No. VCX500220
  • ONLINE EXCLUSIVE PRICE
    58910.00 NOK
    54856.73 NOK / Pack of 1
    Save 4053.27 NOK
    6% Off
View all promotions
Estimated Shipment: 21-01-2027
Safely processes a wide range of organic and inorganic materials, from 250 microliters to 1 liter.
  • Ultrasonic processors for small and medium volume applications
  • Safely processes a wide range of organic and inorganic materials, from 250 microliters to 1 liter 
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical processing
  • Energy and temperature set-point control, digital wattmeter, automatic tuning, and 10 hour process timer
Specifications
Wattage 500 W
Dimensions (L x W x H) 13.5 x 7.5 x 9.25 in. (340 x 190 x 235 mm)
For Use With (Application) Nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical processing
Includes Standard 0.5 in. diameter probe with replaceable tip
Timer 10 hour process timer