Learn More
Sonics Materials VC-505 Ultrasonic Processor
Description
- Extremely capable and versatile, it can safely process a wide range of organic and inorganic materials - from microliters to liters
- 500 and 750 Watt Ultrasonic Processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
- Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
- Energy monitor, digital wattmeter, microprocessor-based and programmable
- 10 hour process timer
Specifications
Specifications
| For Use With (Application) | Processing a wide range of organic and inorganic materials, from 250 uL to 1 L |
| Includes | 0.5 in. (13 mm) probe with replaceable tip, tool kit, and instruction manual |
| Timer | 10 hr. process timer |