missing translation for 'onlineSavingsMsg'
Learn More

VAV2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21924-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891475

  • 1611.84 NOK / 20µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

VAV2
Polyclonal
Unconjugated
VAV2
VAV 2, VAV2
Rabbit
Antigen Affinity Chromatography
RUO
22325, 296603, 7410
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52735, Q60992
VAV2
VAV2 Fusion Protein Ag16458
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers