Learn More
Vaspin Mouse anti-Human, Unconjugated, Clone: 2A1B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67573-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The Vaspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Vaspin. This antibody recognizes human antigen. The Vaspin antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Vaspin | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2A1B6 | |
Unconjugated | |
150 μL | |
145264 | |
IgG1 |