Learn More
VACUUBRAND™ VARIO™ Chemistry Pumping Unit, PC 3004 VARIO Select
GREENER_CHOICE
Product Code. 16251792
Change view
Click to view available options
Plug Type:
CEE
Kaltgerätestecker EN 60320
Model:
PC 3004 VARIO Select
PC 3004 VARIO Select EKP
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Model unitSize
16251792 Kaltgerätestecker EN 60320 PC 3004 VARIO Select EKP Each
15950231 CEE PC 3004 VARIO Select Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16251792 Supplier VACUUBRAND™ Supplier No. 20737574
Provides pinpoint vacuum control by adjusting the motor speed, an exhaust vapor condenser allows for near complete recovery of solvents. The automatic evaporation mode is perfect for controlling applications such as rotary evaporation.
  • VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Powerful vacuum pump meets high vacuum requirements for most high boiling solvents
  • Extraordinary diaphragm life for minimum operational and servicing costs
  • Excellent environmental compatibility through low energy consumption and efficient solvent recovery
  • Pump is suitable for demanding applications, even with high boiling point solvents
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • VACUU·SELECT Controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control during solvent evaporation
  • VARIO motor speed control for exceptionally long maintenance intervals
  • Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Exhaust vapor condenser allows for near complete recovery of solvents when used with a chilled water line or recirculating chiller
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 1.5 Mbar (Without Gas Ballast), 3 Mbar (With Gas Ballast)
Control VACUU·SELECT
Shipping Weight (Metric) 30.5 kg
Includes Pumping Unit with Peltronic™ Emission Condenser with Manual
Motor Speed 30 to 2400 min-1
Certifications/Compliance CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Dimensions (L x W x H) 419 x 243 x 457 mm
Description Peltronic™ Emission Condenser, without Cable
Material PTFE (Partially Carbon Reinforced), ETFE/ECTFE (Partially Carbon Fiber Reinforced), FFKM, Ceramics, PPS
Model PC 3004 VARIO Select EKP
Noise Level 43 dBA
Plug Type Kaltgerätestecker EN 60320
Wattage 0.53 kW
Weight (Metric) 25.2 kg
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 4.6 m³/hr.
Frequency 50-60 Hz
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 4
Applications Intelligent Vacuum with VARIO™-control, Rotary Evaporator/Parallel Evaporator, Vacuum Concentrators, Drying Chambers
No. of Stages 3
Maximum Pressure 1.1 bar
Voltage 200/230 V