Learn More
VACUUBRAND™ Chemistry Pumping Unit, PC 3001 VARIO select
GREENER_CHOICE
Product Code. 16251762
Click to view available options
Plug Type:
CEE
IEC
Model:
PC 3001 VARIO Select
PC 3001 VARIO Select EK Peltronic
PC 3001 VARIO Select IK
PC 3001 VARIO Select TE
Unit Size:
Each
Product Code. 16251762
Brand: VACUUBRAND™ 20700220
Only null left
Add to basket
Add to basket
Provides pinpoint vacuum control by adjusting the motor speed. Automatic evaporation mode is perfect for controlling applications such as rotary evaporation. Various condenser types are available to meet a range of desired applications.
  • VACUU·SELECT controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control for solvent evaporation
  • Simplify lab work with a graphic user interface and predefined applications
  • Automatic boiling point detection and continuous optimization of vacuum levels for quick process times
  • Compact and powerful; superior performance even with continuous condensate purge
  • Extremely low power consumption, efficient solvent recovery, and oil-free for eco-friendly operation
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • VARIO motor speed control for exceptionally long maintenance intervals
  • Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Suitable for use with high boiling point solvents
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
  • Whisper quiet and ultra low vibration
TRUSTED_SUSTAINABILITY
Tekniske data
Max. Vacuum 2 Mbar (Without Gas Ballast), 4 Mbar (With Gas Ballast)
Control VACUU·SELECT
Shipping Weight (Metric) 12.9 kg
Includes Ready-to-Use Pump (Completely Mounted with Manual), Dry Ice Condenser
Motor Speed 200 to 3000 min-1
Certifications/Compliance CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Electrical Requirements 200/230 V, 50-60 Hz and 100/120 V, 50-60 Hz
Dimensions (L x W x H) 301 x 341 x 493 mm
Description Dry ice condenser eliminates the need for a cooling water connection
Material PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
Model PC 3001 VARIO Select TE
Noise Level 42 dBA
Plug Type CEE
Wattage 0.16 kW
Weight (Metric) 9.7 kg
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 2 m³/hr.
Frequency 50-60 Hz
Pump Type Diaphragm Pump
Enclosure Rating IP20
No. of Pump Heads 4
No. of Stages 3
Maximum Pressure 1.1 bar
Voltage 200/230 V
Vis mere Vis mindre
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.