Learn More
Biochrom Variable Pathlength Cell Holder
Product Code. 11603024 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Cell Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11603024 Cell Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11603024 Supplier Biochrom Supplier No. 80710005
  • ONLINE EXCLUSIVE PRICE
    12985.00 NOK
    11923.31 NOK / Pack of 1
    Save 1061.69 NOK
    8% Off
Estimated Shipment: 13-01-2027
Variable Pathlength Cell Holder for use with Ultrospec 7000/8000/9000.
  • Holds 1, 2, 5, 10, 20, 50 and 100 mm pathlength cells
  • Includes one thumb screw
Specifications
Type Cell Holder
Disposable Reusable
For Use With (Equipment) Ultrospec 7000/8000/9000
Holds 1, 2, 5, 10, 20, 50 and 100 mm Pathlength Cells
Includes 1 x Thumb Screw
Shape Rectangular