missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-98897-0.025ML
This item is not returnable.
View return policy
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Vanilloid R1/TRPV1 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TRPV1. | |
| Human, Mouse, Rat | |
| Antisera |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000 | |
| Q8NER1 | |
| TRPV1 | |
| A synthetic peptide from the 3d cytoplasmic loop of human, rat, mouse TRPV1 conjugated to an immunogenic carrier protein was used as the immunogen. | |
| 0.025 mL | |
| Neuroscience, Sensory Systems | |
| 7442 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. Add glycerol (1:1) for additional stability. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction