missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97417JF669
This item is not returnable.
View return policy
Description
Vanilloid R1/TRPV1 Polyclonal antibody specifically detects Vanilloid R1/TRPV1 in Human,Invertebrate,Mouse,Primate,Rat samples. It is validated for Western Blot,Immunohistochemistry (Negative),Immunofluorescence,Immunocytochemistry/ Immunofluorescence
Specifications
| Vanilloid R1/TRPV1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Invertebrate, Mouse, Primate, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Negative), Immunofluorescence, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
| A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] | |
| 0.1 mL | |
| Neuroendocrinology, Neuroscience, Phospho Specific, Sensory Systems | |
| 7442 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction