Learn More
Thermo Scientific Chemicals Vandetanib
Product Code. p-7214490 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 16594404
Brand: Thermo Scientific Chemicals 464332500
Request Bulk or Custom Format
CAS: 443913-73-3 | C22H24BrFN4O2 | 475.36 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 443913-73-3
Molecular Formula C22H24BrFN4O2
Molecular Weight (g/mol) 475.36
InChI Key UHTHHESEBZOYNR-UHFFFAOYSA-N
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
SMILES COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1
Specifications
Melting Point 240.0°C to 243.0°C
Solubility Soluble in methanol
Quantity 250 mg
Formula Weight 475.35
Chemical Name or Material Vandetanib