Learn More
Thermo Scientific Chemicals Vandetanib
Product Code. 16574404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
16574404 1 g 1g
16594404 250 mg 250mg
16584404 5 g 5g
3 options
Product Code. 16574404
Brand: Thermo Scientific Chemicals 464330010
Request Bulk or Custom Format
CAS: 443913-73-3 | C22H24BrFN4O2 | 475.36 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 443913-73-3
Molecular Formula C22H24BrFN4O2
Molecular Weight (g/mol) 475.36
InChI Key UHTHHESEBZOYNR-UHFFFAOYSA-N
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
SMILES COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1
Specifications
Melting Point 240.0°C to 243.0°C
Solubility Soluble in methanol
Quantity 1 g
Formula Weight 475.35
Chemical Name or Material Vandetanib