Learn More
Valtrate, TRC
Specifications
Specifications
| Chemical Name or Material | Valtrate |
| CAS | 18296-44-1 |
| Molecular Formula | C22 H30 O8 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C22H30O8/c1-12(2)6-18(24)29-17-8-16-15(9-26-14(5)23)10-27-21(20(16)22(17)11-28-22)30-19(25)7-13(3)4/h8,10,12-13,17,20-21H,6-7,9,11H2,1-5H3/t17-,20+,21-,22+/m0/s1 |
| SMILES | CC(C)CC(=O)O[C@@H]1OC=C(COC(=O)C)C2=C[C@H](OC(=O)CC(C)C)[C@]3(CO3)[C@@H]12 |
| IUPAC Name | [(1S,6S,7R,7aS)-4-(acetyloxymethyl)-1-(3-methylbutanoyloxy)spiro[6,7a-dihydro-1H-cyclopenta[c]pyran-7,2'-oxirane]-6-yl] 3-methylbutanoate |
| Molecular Weight (g/mol) | 422.47 |
| Formula Weight | 422.1941 |