Learn More
OHAUS™ Valor™ 2000 Scale
Shop All Ohaus Products
Click to view available options
Model:
V22PWE15T
V22PWE3T
V22PWE6T
V22XWE1501T
V22XWE15T
V22XWE3T
V22XWE6T
Unit Size:
1 unit
Description
- The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
- A unique IP68 flow-thru design ensures that the NSF-certified Valor 2000 will drain moisture in the scale when used in wet conditions.
- Stabilization time is less than a half-second.
- Large front and rear LED displays allow for easy viewing so two operators can share one scale.
Specifications
Specifications
| Battery Life | 50 Hours with Rechargeable Battery |
| Construction | ABS housing with stainless steel platform (PW), stainless steel housing and platform (XW), up-front level indicator |
| Capacity | 6.00 kg |
| Display Type | Dual LED |
| Readability | 1.000000 g |
| Linearity | 1.000000 g |
| Model | V22XWE6T |
| Protection Class | IPX8 |
| Repeatability | 1.000000 g |
| Stabilization Time | 0.500 sec. |
| Show More |