Learn More
Valnoctamide, MedChemExpress
Product Code. 16475008
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16475008
Brand: Medchem Express HY121877
MedChemExpress Valnoctamide (Valmethamide), a derivative of valproate, suppresses benzodiazepine-refractory status epilepticus. Valnoctamide (Valmethamide) acts directly on GABAA receptors.
Valnoctamide (Valmethamide), a derivative of valproate, suppresses benzodiazepine-refractory status epilepticus. Valnoctamide (Valmethamide) acts directly on GABAA receptors.
Specifications
Chemical Name or Material Valnoctamide
Physical Form Solid
CAS 4171-13-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H17NO
For Use With (Application) Neuroscience-Neuromodulation
Synonym Valmethamide
Solubility Information DMSO : ≥ 100 mg/mL (698.18 mM)
SMILES CCC(C(C(N)=O)CC)C
Molecular Weight (g/mol) 143.23
Formula Weight 143.23
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.