Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Product Code. 10423305 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Ferrule
Male Nut
Two-piece Fingerlok Nut with PEEK Ferrule
Valco 10-32 male hex nut
Quantity:
1 Each
10 Pack
Unit Size:
Each
Pack of 10
Product Code. Description Quantity unitSize
10423305 Valco 10-32 male hex nut 10 Pack Pack of 10
10639865 Two-piece Fingerlok Nut with PEEK Ferrule 1 Each Each
10446934 Male Nut 10 Pack Pack of 10
10170983 Ferrule 10 Pack Pack of 10
4 options
Product Code. 10423305
Brand: Thermo Scientific™ U320X
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
  • Designed for 10-32 port configurations
  • Burr and contaminant free
TRUSTED_SUSTAINABILITY
Specifications
Type High Pressure Valco Male Hex Nut
Description Valco 10-32 male hex nut
For Use With (Equipment) HPLC Injectors with 10-32 Port Configuration
Material Stainless Steel
Quantity 10 Pack