Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Shop All Thermo Scientific ProductsDescription
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
- Designed for 10-32 port configurations
- Burr and contaminant free
Specifications
Specifications
| Type | Universal Ferrule |
| Description | Ferrule |
| Size (English) | 0.625 in. |
| For Use With (Equipment) | 1/16” tubing |
| Material | Stainless Steel |
| Quantity | 10 Pack |