missing translation for 'onlineSavingsMsg'
Learn More

Val-Cit-PAB-MMAE, MedChemExpress

Product Code. p-200100730
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. Quantity unitSize
16440639 1 mg 1mg
16435717 5 mg 5mg
16412978 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16440639

Brand: Medchem Express HY100374

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Val-Cit-PAB-MMAE is a drug-linker conjugate for ADC. Val-Cit-PAB-MMAE contains the ADCs linker (peptide Val-Cit-PAB) and a potent tubulin inhibitor MMAE (HY-15162). MMAE a potent mitotic inhibitor by inhibiting tubulin polymerization.

Val-Cit-PAB-MMAE is a drug-linker conjugate for ADC. Val-Cit-PAB-MMAE contains the ADCs linker (peptide Val-Cit-PAB) and a potent tubulin inhibitor MMAE (HY-15162). MMAE a potent mitotic inhibitor by inhibiting tubulin polymerization.

Specifications

Chemical Name or Material Val-Cit-PAB-MMAE
Physical Form Solid
CAS 644981-35-1
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C58H94N10O12
Solubility Information DMSO : 50 mg/mL (44.51 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@H](C(C)C)NC([C@H](C(C)C)N(C(OCC3=CC=C(NC([C@H](CCCNC(N)=O)NC([C@H](C(C)C)N)=O)=O)C=C3)=O)C)=O)=O)C
Molecular Weight (g/mol) 1123.43
Formula Weight 1123.43
Percent Purity 95.73%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.