Learn More
Thermo Scientific™ Vacuum Tap for Rear Wall
Product Code. 10756344 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Laboratory Enclosure
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10756344 Laboratory Enclosure Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10756344 Supplier Thermo Scientific™ Supplier No. 50068232
  • ONLINE EXCLUSIVE PRICE
    3105.00 NOK
    2784.31 NOK / Pack of 1
    Save 320.69 NOK
    10% Off
 Restricted Item
In Stock
For Herasafe 2025, 2030i and Maxisafe 2030i Biological Safety Cabinet
For Herasafe 2025, 2030i and Maxisafe 2030i Biological Safety Cabinet
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Laboratory Enclosure
Product Type Add-A-Valve Kit for Main Chamber