Learn More
Edwards Vacuum Rotary Vane Vacuum Pump Exhaust Oil Mist Filters
Product Code. 12705058
Change view
Click to view available options
For Use With:
RV12, E2M18 pump
RV3, RV5, RV8 Vacuum Pump
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12705058 RV3, RV5, RV8 Vacuum Pump Each
10034970 RV12, E2M18 pump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12705058 Supplier Edwards Supplier No. A46226000..
Only null left
Add to basket
Add to basket
Includes
Supplied with NW25 clamp, centering ring, O ring, and NW25 to 3/4 in. adapter.
Captures oil mist from pump outlet when used with RV Rotary Vane Pumps
Ejection of oil into the atmosphere during usage of gas ballast or pump high gas throughputs can be prevented.
  • Semi-transparent white bottom half for oil level monitoring
  • Integral pressure relief valve opens if oil element is blocked
  • Neutralize oil mist smell
  • Integral pressure relief valve opens if oil element blocks
  • Efficient at 99.999% DOP test
  • Azide proof
Use optional oil return kits for reduce oil level maintenance and savings in pump oil
Certifications
Meets COSHH and health and safety regulations
Specifications
For Use With RV3, RV5, RV8 Vacuum Pump
Type Oil Mist Filter