Learn More
Fisherbrand™ Vacuum Pump with UK, US, and Schuko Cables
Product Code. 12911151 Shop All Fisherbrand Products
Change view
Click to view available options
Pump Type:
Vacuum Pump
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pump Type unitSize
12911151 Vacuum Pump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12911151
Add to basket
Edge
Add to basket
Quiet in operation with very low vibration. the Fisherbrand™ Vacuum Pump is designed for general duty laboratory applications requiring only rough vacuum or moderate pressure.
  • Compact filtration pump with small footprint
  • Economical to purchase and operate
  • Quiet running oil free pump
  • Lightweight and highly portable
  • DN 6/8 hose nozzle For vacuum filtration.
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 100 mbar
Electrical Requirements 90/260 V 50/60 Hz with Schuko UK and US power leads
Dimensions (L x W x H) 14.4 x 12.7 x 19.6 cm (5.6 x 5 x 7.7 in.)
Weight (English) 2 lb.
Weight (Metric) 2.3 kg
For Use With Schuko, UK And US Vacuum Cables
Pump Type Vacuum Pump
Air Displacement 15 L/min.
Applications Aspiration
Maximum Pressure 2.5 bar