Learn More
Welch Ilmvac™ Vacuum Pump Oil
Product Code. 10777544 Shop All Welch Ilmvac Products
Change view
Click to view available options
Volume (Metric):
5 L
18.9 L
20 L
Unit Size:
20 L
5 L
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10777544 5 L 5 L
12790428 18.9 L Each
10420853 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10777544 Supplier Welch Ilmvac™ Supplier No. 80012001
 Restricted Item
Estimated Shipment: 01-12-2026
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
Generic mineral oil for use in most one and two stage rotary vane pumps.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuum Pump Oil
For Use With (Application) For General Use in Rotary Vane Pumps
Pressure 10-5 mabr vapour pressure
Viscosity 118 cSt viscosity at 40°C
Volume (Metric) 5 L