Learn More
Day-Impex Vacuum-insulated Stainless Steel Dewar Flasks
Product Code. 11783619
Product Code. 11783619 Supplier Day-Impex Supplier No. 320003
  • ONLINE EXCLUSIVE PRICE
    7289.93 NOK
    5911.32 NOK / Pack of 1
    Save 1378.61 NOK
    18% Off
Estimated Shipment: 15-03-2027
Dilvac™ vacuum-insulated flasks are entirely stainless steel. Lightweight and compact in design they are suitable for ultra-low or very high temperature applications. Available in a range of capacities from 0.5 L to 6 L.
  • Vacuum-insulated
  • Lightweight stainless steel
  • Ideal for storage or transportation
  • Suitable for ultra-low and very high temperatures