Learn More
Gardner Denver Welch™ Kit 10 Vacuum pump control package with MRV 100, DN 25 KF
Product Code. 13465778 Shop All Welch Ilmvac Products
Change view
Click to view available options
Size:
DN 25 KF
Unit Size:
Each
Product Code. Size unitSize
13465778 DN 25 KF Each
1 options
Product Code. 13465778
Brand: Gardner Denver Welch™ 330054
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For rotary vane pumps with KF25 flanges. Cost-effective package for controlling vacuum pressure up to 0.001mbar.
Vacuum control package consists of:
  • MRV100 vacuum controller
  • PIZA111cr-GOLD vacuum sensor
  • KF25/16 measuring T-piece
  • 2-way solenoid valve
  • Clamps, centring rings and adapters
  • Electrical connections
TRUSTED_SUSTAINABILITY
Specifications
Description DN 25 KF
For Use With Rotary vane pumps with KF25 flanges
Size DN 25 KF
Type Vacuum Control Kit