Learn More
BD Vacutainer™ Multi-Sample Blood Collection Needles
Product Code. 15446233
Change view
Click to view available options
Gauge:
20 G
21 G
Length (Metric):
25.4 mm
38.1 mm
Color:
Green
Yellow
Unit Size:
Pack of 100
Product Code. Gauge Length (Metric) Color unitSize
15446233 20 G 25.4 mm Yellow Pack of 100
13101563 21 G 38.1 mm Green Pack of 100
2 options
Product Code. 15446233
Brand: BD 360214
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be used with multiple blood collection tubes. BD Vacutainer™ Multi-Sample Blood Collection Needles are sterile, double-ended needles for use with Vacutainer tube holders and vacuum blood collection tubes. Available in 1 in. or 1.5 in. needles of 20, 21 or 22 gauge.
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Sterile double-ended needle with a sleeve covered safety valve
  • Advanced low-angle bevel design and silicone lubricant coating ensures gentle and smooth vein entry
  • Draws blood directly from the vein into vacuum collection tubes to prevent exposure
  • Can be used with multiple collection tubes after venipuncture
  • Designed for use with BD Vacutainer™ tube holders and vacuum blood collection tubes
  • Luer adapter (MPN 367300) available separately for use with a catheter or hypodermic needle
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-sample Blood Collection Needle
Sterility Sterile
Certifications/Compliance CE marked
Color Yellow
Connectors Double-sided
Disposable Single-use
For Use With (Application) Collection of Blood Samples
For Use With (Equipment) Vacutainer Blood Collection Tubes, Tube Holder
Gauge 20 G
Hub Material Polystyrene
Latex Free Yes
Length (English) 1 in.
Length (Metric) 25.4 mm
Style Low-angle Bevel
Thickness (Wall) Thin Wall